Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166419.1 | NP_001159891.1 | 4 | 669 | ||
NM_001166420.1 | NP_001159892.1 | 4 | 669 | |||
NM_001166422.1 | NP_001159894.1 | 4 | 669 | |||
NM_018486.2 | NP_060956.1 | 4 | 669 | |||
XM_011530986.2 | XP_011529288.2 | 4 | 664 | |||
XM_011530987.2 | XP_011529289.2 | 4 | 664 | |||
XM_017029640.1 | XP_016885129.1 | 4 | 664 | |||
XM_017029641.1 | XP_016885130.1 | 4 | 664 | |||
XM_017029643.1 | XP_016885132.1 | 4 | 664 | |||
XM_017029645.1 | XP_016885134.1 | 4 | 664 | |||
XM_017029646.1 | XP_016885135.1 | 3 | 213 | |||
XM_017029647.1 | XP_016885136.1 | 4 | 664 | |||
GenBank mRNA | AF212246.1 | - | 4 | 385 | ||
AF230097.1 | - | 4 | 382 | |||
AF245664.1 | - | 4 | 336 | |||
AJ277724.1 | - | 4 | 369 | |||
AK074326.1 | - | 4 | 614 | |||
AK300895.1 | - | 4 | 366 | |||
AY429539.1 | - | 3 | 213 | |||
BC050433.1 | - | 4 | 360 | 5761745 | ||
BQ189619.1 | - | 4 | 419 |