Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018358.2 | NP_060828.2 | 17, 18 | 1799 | ||
XM_011512959.2 | XP_011511261.1 | 17, 18 | 1803 | |||
GenBank mRNA | AK002060.1 | - | 17, 18 | 1776 | ||
AK024758.1 | - | 11, 12 | 998 | |||
AK092415.1 | - | 17, 18 | 1682 | |||
AK097073.1 | - | 16, 17 | 2066 | |||
AK126150.1 | - | 3 | 1559 | |||
AK290106.1 | - | 17, 18 | 1671 | |||
AK299433.1 | - | 17, 18 | 1563 | |||
BC009253.2 | - | 17, 18 | 1678 | 3028775 | ||
BC035652.1 | - | 17, 18 | 1673 | |||
BC051754.2 | - | 17, 18 | 1673 | 6064628 | ||
BC051884.1 | - | 17, 18 | 1673 | 5105761 | ||
KU178694.1 | - | 17, 18 | 1666 | |||
KU178695.1 | - | 17, 18 | 1623 |