Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018359.3 | NP_060829.2 | 5 | 543 | ||
NR_028085.1 | 5 | 518 | ||||
GenBank mRNA | AK002062.1 | - | 5 | 472 | ||
AK091635.1 | - | 5 | 511 | |||
BC010493.1 | - | 4 | 439 | 3048997 | ||
BP264462.1 | - | 5 | 539 | |||
CR457278.1 | - | 4 | 426 | |||
DA537391.1 | - | 5 | 537 |