Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018364.4 | NP_060834.2 | 2 | 1129 | ||
NR_130896.1 | 2 | 1129 | ||||
XM_017001518.1 | XP_016857007.1 | 2 | 1129 | |||
GenBank mRNA | AK002082.1 | - | 2 | 944 | ||
AK022166.1 | - | 1 | 308 | |||
AK292552.1 | - | 2 | 1129 | |||
BC026155.1 | - | 2 | 951 | 4827655 | ||
CR627402.1 | - | 3 | 722 |