Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL22L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099645.1 | NP_001093115.1 | 4 | 1421 | ||
NM_001320451.1 | NP_001307380.1 | 4 | 1418 | |||
NR_135259.1 | 4 | 1488 |