Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021491.2 | 6 | 2623 | |||
NR_109820.1 | 5 | 2584 | ||||
NR_109821.1 | 7 | 2744 | ||||
NR_109822.1 | 3 | 3034 | ||||
NR_109823.1 | 5 | 2581 | ||||
NR_109824.1 | 6 | 2680 | ||||
NR_109825.1 | 9 | 3546 | ||||
GenBank mRNA | AK124699.1 | - | 5 | 3309 |