Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124699.1 | - | 1 | 332 | ||
BC144330.1 | - | 1 | 882 | |||
BC144331.1 | - | 1 | 882 |