Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000447.2 | NP_000438.2 | 13 | 1815 | ||
NM_012486.2 | NP_036618.2 | 13 | 1812 | |||
XM_005273199.3 | XP_005273256.1 | 12 | 1628 | |||
XM_017001835.1 | XP_016857324.1 | 12 | 2136 | |||
XM_017001836.1 | XP_016857325.1 | 12 | 1625 | |||
GenBank mRNA | AK292299.1 | - | 14 | 1764 | ||
BC006365.2 | - | 14 | 2009 | 4131408 | ||
L43964.1 | - | 12 | 1755 | |||
L44577.1 | - | 13 | 1753 |