Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003367.3 | 9 | 1863 | |||
GenBank mRNA | BC041065.1 | - | 8 | 1622 | ||
BX648921.1 | - | 1 | 5101 |