Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003367.3 | 6, 7 | 1336 | |||
GenBank mRNA | BC041065.1 | - | 5, 6 | 1095 | ||
BX648921.1 | - | 1 | 450 |