Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101930085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136255.1 | 1 | 1176 | |||
GenBank mRNA | AK055594.1 | - | 1 | 1168 | ||
BC035153.1 | - | 1 | 1149 | |||
BC049371.1 | - | 1 | 1143 |
Gene Symbol: LINC01239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038977.1 | 6 | 1765 | |||
GenBank mRNA | AK092601.1 | - | 6 | 1766 |