Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039210.4 | NP_001034299.3 | 4 | 1197 | ||
NM_001168385.2 | NP_001161857.1 | 5 | 1294 | |||
NM_001257235.2 | NP_001244164.1 | 4 | 1225 | |||
NM_001257239.2 | NP_001244168.1 | 4 | 1244 | |||
NM_001257240.2 | NP_001244169.1 | 4 | 1329 | |||
NM_001257241.2 | NP_001244170.1 | 4 | 1270 | |||
NM_001324290.1 | NP_001311219.1 | 4 | 1262 | |||
NM_001324291.1 | NP_001311220.1 | 4 | 1152 | |||
NM_001324294.1 | NP_001311223.1 | 4 | 1140 | |||
NM_018466.5 | NP_060936.1 | 4 | 1256 | |||
NR_033125.2 | 5 | 1190 | ||||
GenBank mRNA | AK056347.1 | - | 1 | 250 | ||
BC005336.1 | - | 4 | 1189 | 3996922 |