Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018467.3 | NP_060937.1 | 7, 8 | 656 | ||
XM_017026975.1 | XP_016882464.1 | 8, 9 | 1366 | |||
XM_017026976.1 | XP_016882465.1 | 6, 7 | 606 | |||
GenBank mRNA | AB097050.1 | - | 7, 8 | 627 | ||
AF220052.1 | - | 7, 8 | 627 | |||
BC008455.1 | - | 7, 8 | 604 | 4247241 |