Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288732.1 | NP_001275661.1 | 12 | 4414 | ||
NM_001288733.1 | NP_001275662.1 | 12 | 4406 | |||
NM_018469.4 | NP_060939.3 | 12 | 4435 | |||
XM_011524998.1 | XP_011523300.1 | 12 | 4424 | |||
XM_011524999.1 | XP_011523301.1 | 12 | 4348 | |||
XM_011525000.1 | XP_011523302.1 | 12 | 4399 | |||
XM_017024846.1 | XP_016880335.1 | 12 | 4378 | |||
GenBank mRNA | AB051525.2 | - | 11 | 4233 | ||
AF163260.1 | - | 10 | 2452 | |||
AF220182.1 | - | 8 | 1966 | |||
AK000369.1 | - | 7 | 1740 | |||
AK000758.1 | - | 11 | 2650 | |||
AK091156.1 | - | 11 | 3403 | |||
AL832371.1 | - | 12 | 4314 | |||
AL834251.2 | - | 12 | 4325 | |||
BC008896.2 | - | 11 | 3590 | |||
BC033661.1 | - | 9 | 2117 | |||
BC036672.1 | - | 12 | 4385 | |||
BC040521.1 | - | 12 | 4339 | 4837394 | ||
BC040522.1 | - | 12 | 4387 | |||
BC108279.1 | - | 10 | 2472 | |||
BX648260.1 | - | 3 | 4445 | |||
CR627433.1 | - | 12 | 4406 |