Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024452.2 | NP_001019623.2 | 6 | 1113 | ||
NM_001172285.1 | NP_001165756.1 | 6 | 1113 | |||
GenBank mRNA | AK309010.1 | - | 6 | 1113 |