Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163022.1 | NP_001156494.1 | 7, 8 | 1582 | ||
NM_001163023.1 | NP_001156495.1 | 6, 7 | 1492 | |||
NM_001276389.1 | NP_001263318.1 | 6, 7 | 1519 | |||
NM_018474.4 | NP_060944.3 | 8, 9 | 1745 | |||
XM_011529296.2 | XP_011527598.1 | 8, 9 | 1748 | |||
XM_011529297.2 | XP_011527599.1 | 8, 9 | 1683 | |||
XM_011529298.2 | XP_011527600.1 | 8, 9 | 1676 | |||
XM_011529299.2 | XP_011527601.1 | 7, 8 | 1577 | |||
XM_017027951.1 | XP_016883440.1 | 9, 10 | 1691 | |||
XM_017027952.1 | XP_016883441.1 | 7, 8 | 1523 | |||
XM_017027953.1 | XP_016883442.1 | 9, 10 | 1763 | |||
XM_017027954.1 | XP_016883443.1 | 7, 8 | 1523 | |||
GenBank mRNA | AF220187.1 | - | 4, 5 | 777 | ||
AF451990.1 | - | 8, 9 | 1722 | |||
AK025476.1 | - | 8, 9 | 1676 | |||
AK293166.1 | - | 6, 7 | 1423 | |||
AK293567.1 | - | 6, 7 | 1463 | |||
AK295355.1 | - | 7, 8 | 1482 | |||
AK296399.1 | - | 4, 5 | 1011 | |||
AL110196.1 | - | 4, 5 | 970 | |||
BC039296.1 | - | 8, 9 | 1706 | |||
BC045826.1 | - | 7, 8 | 1469 | 5262165 | ||
BC065550.1 | - | 6, 7 | 1326 | |||
BC090879.2 | - | 4, 5 | 1150 | |||
BC105093.1 | - | 8, 9 | 1648 | 8144096 | ||
BC113370.1 | - | 8, 9 | 1648 | 8322422 |