Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300980.1 | NP_001287909.1 | 12 | 1567 | ||
NM_001300982.1 | NP_001287911.1 | 10 | 1256 | |||
NM_018502.4 | NP_060972.3 | 12 | 1549 | |||
XM_005268477.1 | XP_005268534.1 | 12 | 1552 | |||
XM_011537665.2 | XP_011535967.1 | 11 | 1631 | |||
XM_011537668.2 | XP_011535970.1 | 10 | 1304 | |||
XM_017009617.1 | XP_016865106.1 | 11 | 1793 | |||
XM_017009618.1 | XP_016865107.1 | 11 | 1399 | |||
XM_017009619.1 | XP_016865108.1 | 11 | 1355 | |||
XM_017009620.1 | XP_016865109.1 | 11 | 1340 | |||
GenBank mRNA | AK097088.1 | - | 12 | 1513 | ||
AK289495.1 | - | 10 | 1214 | |||
BC001910.1 | - | 12 | 1490 | 3538001 | ||
BC009618.1 | - | 7 | 782 | 3901182 |