Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100130520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324073.1 | NP_001311002.1 | 2 | 244 | ||
NM_001324076.1 | NP_001311005.1 | 2 | 244 | |||
XM_006722261.3 | XP_006722324.1 | 2 | 347 | |||
XM_011525569.2 | XP_011523871.1 | 2 | 413 | |||
GenBank mRNA | LC013475.1 | - | 2 | 244 | ||
LC013476.1 | - | 2 | 244 |