Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039508.1 | NP_001034597.1 | 2 | 333 | ||
NM_018556.3 | NP_061026.2 | 2 | 333 | |||
NM_080816.2 | NP_543006.2 | 2 | 333 | |||
XM_005260749.3 | XP_005260806.1 | 2 | 369 | |||
XM_011529286.2 | XP_011527588.1 | 2 | 380 | |||
XM_011529287.2 | XP_011527589.1 | 2 | 336 | |||
GenBank mRNA | AB042624.1 | - | 2 | 349 | ||
AK292745.1 | - | 2 | 333 | |||
AY748247.1 | - | 2 | 268 | |||
AY748248.1 | - | 2 | 268 | |||
BC020629.1 | - | 2 | 310 | |||
BC064532.1 | - | 2 | 294 | 5588038 | ||
DA008008.1 | - | 2 | 336 |