Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128426.2 | NP_001121898.1 | 1, 2 | 443 | ||
NM_018569.5 | NP_061039.3 | 1, 2 | 443 | |||
NR_136622.1 | 1, 2 | 443 | ||||
XM_017008378.1 | XP_016863867.1 | 1, 2 | 455 | |||
XM_017008379.1 | XP_016863868.1 | 1, 2 | 450 | |||
GenBank mRNA | AK310787.1 | - | 1, 2 | 442 | ||
AK315299.1 | - | 1, 2 | 343 | |||
AL136628.1 | - | 1, 2 | 361 | |||
BC009485.2 | - | 1, 2 | 349 | 3623656 | ||
BM823928.1 | - | 1, 2 | 55 | |||
BX647702.1 | - | 1, 2 | 396 | |||
DA463845.1 | - | 1, 2 | 445 |