Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287491.1 | NP_001274420.1 | 1 | 278 | ||
XM_005264187.3 | XP_005264244.1 | 2 | 798 | |||
XM_011532682.2 | XP_011530984.1 | 2 | 798 | |||
XM_011532683.2 | XP_011530985.1 | 2 | 1107 | |||
XM_011532684.2 | XP_011530986.1 | 2 | 2539 | |||
XM_011532685.2 | XP_011530987.1 | 2 | 798 | |||
XM_011532686.2 | XP_011530988.1 | 2 | 798 | |||
XM_011532688.2 | XP_011530990.1 | 2 | 798 | |||
XM_011532690.1 | XP_011530992.1 | 1 | 107 | |||
XM_017003566.1 | XP_016859055.1 | 2 | 2638 | |||
GenBank mRNA | HQ220209.1 | - | 1 | 278 |