Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278557.1 | NP_001265486.1 | 2 | 207 | ||
NM_001278558.1 | NP_001265487.1 | 2 | 307 | |||
NM_018639.4 | NP_061109.1 | 2 | 307 | |||
XM_017019642.1 | XP_016875131.1 | 2 | 209 | |||
GenBank mRNA | AF163324.1 | - | 1 | 144 | ||
AF229181.1 | - | 2 | 246 | |||
AK294230.1 | - | 1 | 65 | |||
AK295555.1 | - | 2 | 208 | |||
AK298513.1 | - | 2 | 210 | |||
AK310892.1 | - | 2 | 185 | |||
BC015887.2 | - | 2 | 197 | 3910968 |