Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000593.5 | NP_000584.2 | 9 | 2110 | ||
NM_001292022.1 | NP_001278951.1 | 9 | 1370 | |||
GenBank mRNA | AB012644.1 | - | 9 | 1774 | ||
AK123494.1 | - | 3 | 1697 | |||
AK130176.1 | - | 4 | 522 | |||
AK302340.1 | - | 9 | 1341 | |||
AK313762.1 | - | 9 | 2146 | |||
AY523970.1 | - | 9 | 1981 | |||
AY523971.2 | - | 9 | 1981 | |||
BC014081.2 | - | 9 | 2110 | 4304716 | ||
BX648013.1 | - | 9 | 2118 | |||
KU178346.1 | - | 9 | 1955 | |||
KU178347.1 | - | 3 | 500 | |||
L21204.1 | - | 9 | 1775 | |||
L21205.1 | - | 9 | 1775 | |||
L21206.1 | - | 9 | 1775 | |||
L21207.1 | - | 9 | 1775 | |||
L21208.1 | - | 9 | 1775 | |||
X57522.1 | - | 9 | 1985 |