Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242589.2 | NP_001229518.1 | 2 | 176 | ||
NM_001242590.2 | NP_001229519.1 | 2 | 176 | |||
NM_018643.4 | NP_061113.1 | 2 | 176 | |||
NR_136332.1 | 2 | 176 | ||||
XM_006715117.3 | XP_006715180.1 | 2 | 113 | |||
XM_011514696.1 | XP_011512998.1 | 2 | 116 | |||
XM_011514697.1 | XP_011512999.1 | 2 | 113 | |||
XM_017010956.1 | XP_016866445.1 | 2 | 116 | |||
XM_017010957.1 | XP_016866446.1 | 2 | 113 | |||
GenBank mRNA | AF196329.1 | - | 2 | 133 | ||
AF287008.1 | - | 2 | 150 | |||
AK223264.1 | - | 2 | 113 | |||
AK292162.1 | - | 2 | 113 | |||
AK301519.1 | - | 2 | 113 | |||
AK313104.1 | - | 2 | 128 | |||
AY074783.1 | - | 2 | 103 | |||
BC017773.1 | - | 2 | 131 | 4692680 | ||
BI910026.1 | - | 2 | 96 | |||
DC428714.1 | - | 2 | 176 |