Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 678 | |||
NR_110066.1 | 3 | 360 | ||||
NR_110067.1 | 4 | 616 | ||||
NR_110068.1 | 2 | 316 | ||||
NR_110069.1 | 3 | 508 | ||||
NR_110070.1 | 5 | 467 | ||||
GenBank mRNA | AK090593.1 | - | 3 | 242 | ||
BG478902.1 | - | 361 | ||||
BM544796.1 | - | 5 | 488 | |||
DA025114.1 | - | 3 | 242 | |||
DA689880.1 | - | 2 | 196 |