Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL28A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515358.2 | XP_011513660.1 | 1 | 1191 | ||
XM_017012131.1 | XP_016867620.1 | 1 | 1191 |