Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080416.3 | NP_001073885.1 | 6 | 1023 | ||
NM_001144755.2 | NP_001138227.1 | 6 | 1023 | |||
NM_001294282.1 | NP_001281211.1 | 6 | 1023 | |||
XM_011517533.2 | XP_011515835.1 | 6 | 1064 | |||
XM_011517535.2 | XP_011515837.1 | 6 | 1064 | |||
XM_017013455.1 | XP_016868944.1 | 6 | 874 | |||
XM_017013456.1 | XP_016868945.1 | 6 | 871 | |||
XM_017013457.1 | XP_016868946.1 | 6 | 872 | |||
XM_017013458.1 | XP_016868947.1 | 6 | 869 | |||
XM_017013459.1 | XP_016868948.1 | 6 | 876 | |||
GenBank mRNA | BC101186.1 | - | 6 | 681 | 40018967 | |
BC101188.1 | - | 6 | 681 | 40018969 | ||
X13269.1 | - | 6 | 716 | |||
X13294.1 | - | 6 | 716 |