Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242338.2 | NP_001229267.2 | 28 | 6451 | ||
NM_017552.3 | NP_060022.2 | 28 | 6466 | |||
NR_125717.1 | 29 | 6535 | ||||
XM_005264372.3 | XP_005264429.2 | 29 | 6508 | |||
XM_011532921.2 | XP_011531223.1 | 29 | 6493 | |||
GenBank mRNA | AB033066.1 | - | 12 | 3949 | ||
AF038204.1 | - | 1 | 1098 |