Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020700.1 | NP_065751.1 | 10 | 5127 | ||
XM_011538578.2 | XP_011536880.1 | 10 | 5263 | |||
XM_011538579.1 | XP_011536881.1 | 10 | 4815 | |||
GenBank mRNA | AB032983.1 | - | 9 | 4721 | ||
AB084258.1 | - | 10 | 5392 | |||
BC157843.1 | - | 10 | 5139 | 9057062 |