Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199107.1 | NP_001186036.1 | 8 | 4973 | ||
NM_020705.2 | NP_065756.1 | 7 | 4955 | |||
XM_017023493.1 | XP_016878982.1 | 9 | 4278 | |||
XM_017023494.1 | XP_016878983.1 | 7 | 5085 | |||
XM_017023495.1 | XP_016878984.1 | 8 | 4260 | |||
GenBank mRNA | BX648283.1 | - | 4 | 3702 |