Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018686.5 | NP_061156.1 | 2, 3 | 543 | ||
NR_135117.1 | 2, 3 | 543 | ||||
GenBank mRNA | AF271388.1 | - | 2, 3 | 431 | ||
AK022927.1 | - | 2, 3 | 453 | |||
AK222981.1 | - | 2, 3 | 442 | |||
AL832975.1 | - | 2, 3 | 491 | |||
BC016609.1 | - | 2, 3 | 492 | 3448277 |