Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 9 | 823 | ||
XM_005248177.1 | XP_005248234.1 | 8 | 778 | |||
XM_011513872.2 | XP_011512174.1 | 8 | 778 | |||
XM_017008482.1 | XP_016863971.1 | 6 | 572 | |||
GenBank mRNA | AB037766.1 | - | 6 | 561 | ||
AK096047.1 | - | 8 | 765 | |||
AK309178.1 | - | 8 | 753 | |||
BC103710.1 | - | 6 | 551 | |||
BX647334.1 | - | 8 | 758 | |||
DB043556.1 | - | 6 | 572 | |||
EU450799.1 | - | 9 | 650 |