Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020823.1 | NP_065874.1 | 17 | 4292 | ||
XM_005267074.3 | XP_005267131.1 | 17 | 4009 | |||
XM_011535999.1 | XP_011534301.1 | 17 | 4320 | |||
XM_011536000.1 | XP_011534302.1 | 15 | 4147 | |||
XM_011536001.2 | XP_011534303.1 | 17 | 4500 | |||
XM_011536002.2 | XP_011534304.1 | 17 | 4474 | |||
GenBank mRNA | AB037844.1 | - | 17 | 4292 | ||
AJ420476.1 | - | 1 | 802 | |||
AJ420523.1 | - | 1 | 963 | |||
AL359569.1 | - | 1 | 1260 | |||
BC033450.1 | - | 16 | 4015 | |||
BC044210.1 | - | 1 | 887 | |||
BC046100.1 | - | 1 | 384 | |||
BC058328.1 | - | 1 | 646 | |||
BC067769.1 | - | 1 | 362 | |||
BC092402.1 | - | 1 | 372 | |||
BC108681.1 | - | 1 | 654 |