Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020846.1 | NP_065897.1 | 8 | 2015 | ||
XM_005248121.2 | XP_005248178.1 | 9 | 2209 | |||
GenBank mRNA | AB040891.1 | - | 8 | 1985 | ||
AK026476.1 | - | 8 | 1563 | |||
AK291006.1 | - | 8 | 2145 | |||
BC031691.2 | - | 6 | 1335 | |||
BC040993.1 | - | 8 | 2294 | 5272534 |