Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020883.1 | NP_065934.1 | 10 | 2495 | ||
XM_011541861.2 | XP_011540163.1 | 10 | 2503 | |||
XM_017001913.1 | XP_016857402.1 | 8 | 1404 | |||
GenBank mRNA | AB040944.1 | - | 10 | 2473 | ||
BC013836.1 | - | 4 | 646 |