Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 19 | 4066 | ||
NM_020902.1 | NP_065953.1 | 17 | 3985 | |||
XM_005272491.3 | XP_005272548.1 | 18 | 4002 | |||
XM_011528154.2 | XP_011526456.1 | 19 | 3985 | |||
GenBank mRNA | AF289580.1 | - | 7 | 1938 |