Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127208.2 | NP_001120680.1 | 10 | 4830 | ||
XM_005263082.2 | XP_005263139.1 | 9 | 4706 | |||
XM_006714242.3 | XP_006714305.1 | 8 | 4374 | |||
GenBank mRNA | AB046766.1 | - | 3 | 390 | ||
AB075496.1 | - | 7 | 1884 | |||
BC110509.1 | - | 5 | 754 | |||
BC110510.2 | - | 5 | 754 | |||
FM992369.1 | - | 10 | 5203 |