Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 2 | 7853 | ||
XM_011516246.2 | XP_011514548.1 | 3 | 8182 | |||
XM_011516247.2 | XP_011514549.1 | 3 | 8132 | |||
XM_011516249.2 | XP_011514551.1 | 3 | 7983 | |||
XM_011516250.2 | XP_011514552.1 | 3 | 7471 | |||
XM_017012231.1 | XP_016867720.1 | 3 | 7679 | |||
GenBank mRNA | AB038784.1 | - | 1 | 310 | ||
AF007194.1 | - | 3 | 1331 |