Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021214.1 | NP_067037.1 | 3 | 1914 | ||
GenBank mRNA | AK091780.1 | - | 1 | 1891 | ||
AK092758.1 | - | 3 | 1993 | |||
AL390079.1 | - | 3 | 1535 | |||
BC059401.1 | - | 3 | 1884 | 30346383 | ||
BC115003.1 | - | 3 | 1813 | 7503983 |