Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080418.2 | NP_001073887.1 | 11 | 2968 | ||
XM_011541879.2 | XP_011540181.1 | 12 | 4853 | |||
XM_011541880.2 | XP_011540182.1 | 7 | 2677 | |||
GenBank mRNA | AF131778.1 | - | 4 | 739 | ||
AK294907.1 | - | 3 | 312 | |||
BC114370.1 | - | 7 | 712 |