Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098814.1 | NP_001092284.1 | 6 | 1402 | ||
NM_001323667.1 | NP_001310596.1 | 7 | 1532 | |||
NM_001323668.1 | NP_001310597.1 | 7 | 2780 | |||
XM_017023527.1 | XP_016879016.1 | 7 | 2727 | |||
XM_017023528.1 | XP_016879017.1 | 6 | 1433 | |||
XM_017023529.1 | XP_016879018.1 | 6 | 1448 | |||
GenBank mRNA | AK302601.1 | - | 6 | 1438 | ||
AL831973.2 | - | 6 | 1374 | |||
AL832361.1 | - | 7 | 2774 | |||
BX647476.1 | - | 7 | 2774 |