Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316349.1 | NP_001303278.1 | 21 | 3978 | ||
XM_017005049.1 | XP_016860538.1 | 15 | 2455 | |||
GenBank mRNA | AB051466.1 | - | 20 | 3608 |