Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030647.1 | NP_085150.1 | 20 | 5173 | ||
XM_011516587.2 | XP_011514889.1 | 14 | 4198 | |||
GenBank mRNA | AB051505.1 | - | 8 | 3478 | ||
BX641017.1 | - | 13 | 4047 | |||
BX649115.1 | - | 15 | 4439 |