Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206998.1 | NP_001193927.1 | 8 | 1408 | ||
NM_032173.3 | NP_115549.2 | 8 | 1201 | |||
XM_011530435.2 | XP_011528737.1 | 9 | 1397 | |||
XM_011530436.2 | XP_011528738.1 | 10 | 1452 | |||
XM_011530437.1 | XP_011528739.1 | 9 | 2122 | |||
XM_011530438.2 | XP_011528740.1 | 9 | 1638 | |||
XM_017028990.1 | XP_016884479.1 | 9 | 1893 | |||
GenBank mRNA | AB051436.1 | - | 8 | 1273 | ||
AK025710.1 | - | 3 | 595 | |||
AK096397.1 | - | 8 | 1201 | |||
BC009252.2 | - | 3 | 532 | |||
BC021570.1 | - | 3 | 560 | |||
BC069019.1 | - | 3 | 531 | |||
BC094857.1 | - | 3 | 531 | |||
CR456397.1 | - | 8 | 1181 |