Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM161A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201543.1 | NP_001188472.1 | 6 | 2089 | ||
NM_032180.2 | NP_115556.2 | 5 | 1921 | |||
NR_037710.1 | 5 | 1865 | ||||
XM_017005072.1 | XP_016860561.1 | 5 | 1874 | |||
XM_017005073.1 | XP_016860562.1 | 6 | 2016 | |||
XM_017005074.1 | XP_016860563.1 | 5 | 1821 | |||
GenBank mRNA | AK091575.1 | - | 1 | 950 | ||
AK296255.1 | - | 4 | 1455 | |||
BC036448.1 | - | 5 | 1819 | |||
BC107162.1 | - | 5 | 1693 | |||
BC107163.1 | - | 5 | 1693 | |||
BX648834.1 | - | 6 | 1999 | |||
BX649029.1 | - | 5 | 1832 |