Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032184.1 | NP_115560.1 | 7 | 3215 | ||
XM_017002483.1 | XP_016857972.1 | 7 | 3890 | |||
XM_017002485.1 | XP_016857974.1 | 7 | 2602 | |||
XM_017002486.1 | XP_016857975.1 | 7 | 2515 |