Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032184.1 | NP_115560.1 | 4 | 2658 | ||
XM_006710946.3 | XP_006711009.1 | 4 | 3337 | |||
XM_017002483.1 | XP_016857972.1 | 4 | 3338 | |||
XM_017002484.1 | XP_016857973.1 | 4 | 3337 | |||
XM_017002485.1 | XP_016857974.1 | 4 | 2045 | |||
XM_017002486.1 | XP_016857975.1 | 4 | 1958 |