Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032428.1 | NP_115804.1 | 6 | 467 | ||
XM_011531056.2 | XP_011529358.1 | 2 | 159 | |||
XM_011531057.2 | XP_011529359.1 | 2 | 159 | |||
XM_011531058.2 | XP_011529360.1 | 2 | 159 | |||
XM_017029901.1 | XP_016885390.1 | 4 | 688 | |||
XM_017029902.1 | XP_016885391.1 | 4 | 688 |
Gene Symbol: FRMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA661738.1 | - | 2 | 442 |