Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033390.1 | NP_203748.1 | 6 | 2888 | ||
XM_005271715.3 | XP_005271772.1 | 6 | 3269 | |||
XM_011543055.2 | XP_011541357.1 | 6 | 3001 | |||
XM_017018477.1 | XP_016873966.1 | 6 | 7439 | |||
GenBank mRNA | AB051513.1 | - | 4 | 2061 | ||
AB096241.1 | - | 5 | 3625 | |||
AK295437.1 | - | 6 | 2889 | |||
AK299100.1 | - | 6 | 2921 | |||
AL832893.1 | - | 3 | 1741 |