Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024172.1 | 1, 2 | 480 | |||
GenBank mRNA | AK025311.1 | - | 1, 2 | 601 | ||
BC105715.1 | - | 1, 2 | 480 |